siRNA Details
Gene Symbol: Rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169146.1 | NP_001162617.1 | 4, 5 | 368 | ||
XM_006228612.2 | XP_006228674.1 | 3, 4 | 454 | |||
GenBank mRNA | BC064030.1 | - | 342 | |||
BC084715.1 | - | 4, 5 | 337 | |||
BC159438.1 | - | 4, 5 | 339 | |||
FQ211832.1 | - | 4, 5 | 349 | |||
FQ221089.1 | - | 4, 5 | 356 | |||
FQ221637.1 | - | 4, 5 | 350 | |||
FQ221660.1 | - | 4, 5 | 349 | |||
FQ221692.1 | - | 4, 5 | 347 | |||
FQ222913.1 | - | 4, 5 | 349 | |||
FQ223100.1 | - | 4, 5 | 350 | |||
FQ223384.1 | - | 4, 5 | 349 | |||
FQ223396.1 | - | 4, 5 | 350 | |||
FQ224101.1 | - | 4, 5 | 349 | |||
FQ228433.1 | - | 4, 5 | 350 | |||
FQ229060.1 | - | 4, 5 | 302 | |||
X17665.1 | - | 4, 5 | 348 |

More Information
Other Species Targeted

Gene Ontology Categories:
