siRNA Details
Gene Symbol: ABCC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079528.3 | NP_001072996.1 | 2 | 87 | ||
NM_001171.5 | NP_001162.4 | 2 | 87 | |||
XM_011522479.2 | XP_011520781.1 | 2 | 231 | |||
XM_011522482.2 | XP_011520784.1 | 2 | 228 | |||
XM_017023212.1 | XP_016878701.1 | 2 | 231 | |||
XM_017023213.1 | XP_016878702.1 | 2 | 90 | |||
XM_017023214.1 | XP_016878703.1 | 2 | 232 | |||
GenBank mRNA | AF076622.1 | - | 2 | 73 | ||
AF168791.1 | - | 2 | 50 | |||
AK298295.1 | - | 2 | 110 | |||
AM711638.1 | - | 2 | 50 | |||
AM774324.1 | - | 2 | 50 | |||
AY078405.1 | - | 2 | 96 | |||
BC050733.1 | - | 2 | 72 | |||
BC131732.1 | - | 2 | 73 | 40124570 | ||
DC361874.1 | - | 2 | 110 |
Gene Symbol: ABCC6P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003569.1 | 2 | 78 | |||
GenBank mRNA | AK298443.1 | - | 2 | 138 |
Gene Symbol: ABCC6P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023387.1 | 2 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
