siRNA Details
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001680.4 | NP_001671.2 | 5 | 257 | ||
NM_021603.3 | NP_067614.1 | 5 | 264 | |||
GenBank mRNA | AA309757.1 | - | 5 | 252 | ||
AF241236.1 | - | 5 | 232 | |||
BC005302.1 | - | 5 | 264 | 3828677 | ||
BC013289.1 | - | 5 | 260 | 3686762 | ||
BX641053.1 | - | 1 | 1594 | |||
U50743.1 | - | 5 | 221 |
Gene Symbol: FXYD6-FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204268.1 | NP_001191197.1 | 10 | 870 | ||
NM_001243598.2 | NP_001230527.1 | 9 | 808 | |||
GenBank mRNA | CN310658.1 | - | 9 | 443 | ||
FY211982.1 | - | 10 | 467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
