siRNA Details
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001680.4 | NP_001671.2 | 2, 3 | 125 | ||
NM_021603.3 | NP_067614.1 | 2, 3 | 132 | |||
GenBank mRNA | AA309757.1 | - | 2, 3 | 120 | ||
AF241236.1 | - | 2, 3 | 100 | |||
BC005302.1 | - | 2, 3 | 132 | 3828677 | ||
BC013289.1 | - | 2, 3 | 128 | 3686762 | ||
BT006721.1 | - | 2, 3 | 53 | |||
U50743.1 | - | 2, 3 | 89 |
Gene Symbol: FXYD6-FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204268.1 | NP_001191197.1 | 7, 8 | 738 | ||
GenBank mRNA | FY211982.1 | - | 7, 8 | 335 | ||
FY214031.1 | - | 7, 8 | 335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
