siRNA Details
Gene Symbol: BLVRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000713.2 | NP_000704.1 | 5 | 701 | ||
GenBank mRNA | AK312137.1 | - | 5 | 628 | ||
BC109371.1 | - | 5 | 618 | 5229026 | ||
BU528444.1 | - | 3 | 296 | |||
D26308.1 | - | 5 | 649 | |||
D32143.1 | - | 5 | 680 | |||
EU794593.1 | - | 5 | 701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
