siRNA Details
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 5 | 816 | ||
GenBank mRNA | AK291587.1 | - | 5 | 735 | ||
BC002717.2 | - | 5 | 709 | 3631916 | ||
BC005921.1 | - | 5 | 728 | 4043768 | ||
BC020756.1 | - | 5 | 784 | 4769769 | ||
BC057768.1 | - | 5 | 728 | 30337725 | ||
BC062775.1 | - | 5 | 732 | 4604848 | ||
J00118.1 | - | 5 | 695 | |||
M25118.1 | - | 1 | 189 | |||
V00593.1 | - | 4 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
