siRNA Details
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 5 | 750 | ||
NM_022644.3 | NP_072170.1 | 4 | 1003 | |||
NM_022645.2 | NP_072171.1 | 3 | 465 | |||
GenBank mRNA | BC022044.1 | - | 5 | 701 | 4769046 | |
BC035965.1 | - | 5 | 697 | 4731880 | ||
BC119748.2 | - | 5 | 694 | 40119242 | ||
V00573.1 | - | 5 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
