siRNA Details
Gene Symbol: ETV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300974.1 | NP_001287903.1 | 2 | 448 | ||
NM_001304549.1 | NP_001291478.1 | 3 | 545 | |||
NM_014209.3 | NP_055024.2 | 3 | 545 | |||
XM_005258652.2 | XP_005258709.1 | 2 | 609 | |||
XM_011526624.2 | XP_011524926.1 | 2 | 1010 | |||
XM_017026472.1 | XP_016881961.1 | 2 | 609 | |||
GenBank mRNA | AK097439.1 | - | 2 | 609 | ||
BC040908.1 | - | 2 | 392 | |||
BC107123.1 | - | 3 | 143 | |||
BC107124.1 | - | 3 | 143 | |||
BC140746.1 | - | 3 | 235 | 9021614 | ||
BC144448.1 | - | 3 | 236 | 9052974 | ||
DB046527.1 | - | 3 | 537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
