siRNA Details
Gene Symbol: ETV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300974.1 | NP_001287903.1 | 4 | 881 | ||
NM_014209.3 | NP_055024.2 | 5 | 978 | |||
XM_005258652.2 | XP_005258709.1 | 4 | 1042 | |||
XM_011526624.2 | XP_011524926.1 | 4 | 1459 | |||
XM_017026472.1 | XP_016881961.1 | 3 | 961 | |||
GenBank mRNA | AK097439.1 | - | 4 | 1042 | ||
BC040908.1 | - | 4 | 825 | |||
BC140746.1 | - | 5 | 668 | 9021614 | ||
BC144448.1 | - | 5 | 666 | 9052974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
