siRNA Details
Gene Symbol: GAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000805.4 | NP_000796.1 | 2 | 230 | ||
GenBank mRNA | BC069724.1 | - | 2 | 195 | 7262518 | |
BC069762.1 | - | 2 | 195 | 7262530 | ||
BM768483.1 | - | 2 | 229 | |||
V00511.1 | - | 1 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
