siRNA Details
Gene Symbol: GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017998.3 | NP_001017998.1 | 2, 3 | 282 | ||
GenBank mRNA | BC015206.2 | - | 2, 3 | 228 | ||
BC107574.1 | - | 2, 3 | 249 | 4050245 | ||
BG708408.1 | - | 2, 3 | 287 | |||
DB504443.1 | - | 2, 3 | 282 | |||
U31383.1 | - | 2, 3 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
