siRNA Details
Gene Symbol: HIST1H2AD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021065.3 | NP_066409.1 | 1 | 440 | ||
GenBank mRNA | BC093807.1 | - | 1 | 429 | 7939652 | |
BC093809.1 | - | 1 | 432 | 7939654 | ||
BC128035.1 | - | 1 | 390 | 40021702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
