siRNA Details
Gene Symbol: HBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000517.4 | NP_000508.1 | 3 | 490 |
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000558.4 | NP_000549.1 | 3 | 490 | ||
GenBank mRNA | AF097635.1 | - | 3 | 447 | ||
AF105974.1 | - | 3 | 445 | |||
AF147332.1 | - | 3 | 442 | |||
AF281258.1 | - | 2 | 272 | |||
AF349571.1 | - | 3 | 449 | |||
AI064720.1 | - | 2 | 248 | |||
AK223392.1 | - | 3 | 467 | |||
BC005931.1 | - | 3 | 461 | 4047053 | ||
BC008572.1 | - | 3 | 447 | 4177243 | ||
BC032122.2 | - | 3 | 447 | 4849981 | ||
BC050661.1 | - | 3 | 447 | 5209215 | ||
BC101846.1 | - | 3 | 454 | 8069352 | ||
BC101848.1 | - | 3 | 454 | 8069354 | ||
DQ655926.2 | - | 2 | 268 | |||
V00493.1 | - | 3 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
