siRNA Details
Gene Symbol: HLA-DQA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002122.3 | NP_002113.2 | 3 | 419 | ||
XM_006715079.3 | XP_006715142.1 | 3 | 422 | |||
GenBank mRNA | AB209628.1 | - | 3 | 394 | ||
AK130598.1 | - | 3 | 414 | |||
AK130811.1 | - | 3 | 419 | |||
AK130838.1 | - | 3 | 416 | |||
AK313975.1 | - | 3 | 421 | |||
BC008585.1 | - | 3 | 397 | 4339446 | ||
BC051832.1 | - | 4 | 1392 | |||
BC073977.1 | - | 3 | 400 | 6014803 | ||
BC125045.1 | - | 3 | 413 | 40116010 | ||
BC157865.1 | - | 3 | 385 | 9057084 | ||
BC171877.1 | - | 3 | 370 | 9054531 | ||
CR450297.1 | - | 3 | 363 | |||
DQ284436.1 | - | 3 | 428 | |||
DQ284437.1 | - | 3 | 427 | |||
DQ284438.1 | - | 3 | 428 | |||
DQ284439.1 | - | 3 | 427 | |||
DQ284440.1 | - | 3 | 424 | |||
J00199.1 | - | 2 | 250 | |||
JF895477.1 | - | 3 | 366 | |||
L34082.1 | - | 2 | 297 | |||
L34083.1 | - | 2 | 297 | |||
L34084.1 | - | 2 | 297 | |||
L34085.1 | - | 2 | 297 | |||
L34086.1 | - | 2 | 297 | |||
L34087.1 | - | 2 | 294 | |||
L34088.1 | - | 2 | 297 | |||
L34091.1 | - | 2 | 294 | |||
L34092.1 | - | 2 | 294 | |||
L34093.1 | - | 2 | 294 | |||
L42625.1 | - | 2 | 297 | |||
M11124.1 | - | 2 | 297 | |||
M16995.1 | - | 3 | 363 | |||
M17846.1 | - | 1 | 14 | |||
M17847.1 | - | 1 | 14 | |||
M20431.1 | - | 3 | 300 | |||
M20506.1 | - | 2 | 262 | |||
M34996.1 | - | 3 | 330 | |||
U77589.1 | - | 3 | 366 | |||
X00033.1 | - | 3 | 382 | |||
X00370.1 | - | 3 | 373 | |||
X00452.1 | - | 3 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
