siRNA Details
Gene Symbol: RPL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000993.4 | NP_000984.1 | 5 | 462 | ||
NM_001099693.1 | NP_001093163.1 | 4 | 740 | |||
GenBank mRNA | BC017343.2 | - | 4 | 377 | 4895396 | |
BC070210.1 | - | 4 | 676 | 4714258 | ||
BC070373.1 | - | 1 | 398 | 4666695 | ||
BG031488.1 | - | 4 | 379 | |||
BG223187.1 | - | 1 | 416 | |||
CD108396.1 | - | 1 | 370 | |||
CD247800.1 | - | 5 | 431 | |||
CF124259.1 | - | 5 | 456 | |||
X15940.1 | - | 4 | 382 | |||
X69181.1 | - | 4 | 363 |
Gene Symbol: RPL31P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002595.1 | 1 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
