siRNA Details
Gene Symbol: SPRR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006945.4 | NP_008876.3 | 2 | 484 | ||
XM_017002175.1 | XP_016857664.1 | 2 | 516 | |||
GenBank mRNA | AK291317.1 | - | 2 | 486 | ||
M21302.1 | - | 2 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
