siRNA Details
Gene Symbol: TROAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005480.3 | NP_005471.3 | 8 | 1024 | ||
XM_006719181.1 | XP_006719244.1 | 8 | 968 | |||
XM_011537723.1 | XP_011536025.1 | 8 | 968 | |||
XM_011537724.1 | XP_011536026.1 | 7 | 764 | |||
GenBank mRNA | AK296811.1 | - | 8 | 911 | ||
AK299302.1 | - | 8 | 831 | |||
AK309716.1 | - | 5 | 445 | |||
AK310709.1 | - | 7 | 762 | |||
AK315211.1 | - | 8 | 968 | |||
BC032583.1 | - | 8 | 936 | 5498800 | ||
U04810.2 | - | 8 | 967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
