siRNA Details
Gene Symbol: HIST1H3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003536.2 | NP_003527.1 | 1 | 417 | ||
GenBank mRNA | BC007518.2 | - | 1 | 545 | ||
BC096128.3 | - | 1 | 407 | 40002508 | ||
BC096129.3 | - | 1 | 407 | 40002509 | ||
BC096130.3 | - | 1 | 407 | 40002510 | ||
BC143046.1 | - | 1 | 407 | 40112088 | ||
BC148243.1 | - | 1 | 407 | 40082529 | ||
BC148250.1 | - | 1 | 407 | 40002516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
