siRNA Details
Gene Symbol: TROAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005480.3 | NP_005471.3 | 12 | 1369 | ||
XM_006719181.1 | XP_006719244.1 | 12 | 1313 | |||
XM_011537724.1 | XP_011536026.1 | 11 | 1109 | |||
GenBank mRNA | AK315211.1 | - | 12 | 1313 | ||
BC032583.1 | - | 12 | 1281 | 5498800 | ||
U04810.2 | - | 12 | 1312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
