siRNA Details
Gene Symbol: FAM127A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078171.1 | NP_001071639.1 | 1 | 985 | ||
GenBank mRNA | AF038168.1 | - | 1 | 921 | ||
AF052096.1 | - | 1 | 921 | |||
BC002385.2 | - | 1 | 915 | 2961665 | ||
BC002410.2 | - | 1 | 915 | 2961641 | ||
CR456853.1 | - | 1 | 587 | |||
D88756.1 | - | 1 | 587 | |||
Y13374.1 | - | 1 | 930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
