siRNA Details
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF118394.1 | - | 7 | 1295 | ||
AK001023.1 | - | 7 | 1305 | |||
AK093593.1 | - | 5 | 2142 | |||
BC002768.2 | - | 7 | 1286 | 3633393 | ||
BC008005.1 | - | 7 | 1311 | 3507768 | ||
BX648907.1 | - | 5 | 2097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
