siRNA Details
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284501.1 | NP_001271430.1 | 6 | 852 | ||
NM_001284502.1 | NP_001271431.1 | 4 | 463 | |||
NM_012225.3 | NP_036357.1 | 5 | 662 | |||
NR_104318.1 | 4 | 463 | ||||
XM_005255027.2 | XP_005255084.1 | 5 | 1129 | |||
XM_005255030.1 | XP_005255087.1 | 4 | 400 | |||
XM_011522338.2 | XP_011520640.1 | 4 | 597 | |||
XM_017022831.1 | XP_016878320.1 | 5 | 796 | |||
XM_017022832.1 | XP_016878321.1 | 4 | 930 | |||
GenBank mRNA | AF118394.1 | - | 5 | 627 | ||
AK001023.1 | - | 5 | 638 | |||
AK093593.1 | - | 3 | 1475 | |||
AK300696.1 | - | 4 | 396 | |||
AK315611.1 | - | 5 | 603 | |||
BC002768.2 | - | 5 | 619 | 3633393 | ||
BC008005.1 | - | 5 | 644 | 3507768 | ||
BI196883.1 | - | 4 | 388 | |||
BX648907.1 | - | 3 | 1430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
