siRNA Details
Gene Symbol: IFITM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006435.2 | NP_006426.2 | 2 | 482 | ||
GenBank mRNA | BC009696.1 | - | 2 | 426 | 3876542 | |
BG164993.1 | - | 2 | 482 | |||
BG257141.1 | - | 2 | 378 | |||
X02490.1 | - | 2 | 351 | |||
X57351.1 | - | 2 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
