siRNA Details
Gene Symbol: IFITM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC009696.1 | - | 2 | 394 | 3876542 | |
BT009859.1 | - | 2 | 364 | |||
CR456894.1 | - | 2 | 364 | |||
X57351.1 | - | 2 | 643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
