siRNA Details
Gene Symbol: DUX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293798.2 | NP_001280727.1 | 1 | 1170 | ||
NM_001306068.2 | NP_001292997.1 | 1 | 1170 | |||
NR_137167.1 | 1 | 1170 | ||||
GenBank mRNA | HQ266760.1 | - | 1 | 1170 |
Gene Symbol: DBET | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121644.1 | 1 | 1311 | |||
GenBank mRNA | JQ639078.1 | - | 2 | 1309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
