siRNA Details
Gene Symbol: TOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135729.1 | NP_001129201.1 | 13, 14 | 1341 | ||
NM_001135730.1 | NP_001129202.1 | 12, 13 | 1279 | |||
NM_001135732.1 | NP_001129204.1 | 13, 14 | 1414 | |||
NM_005488.2 | NP_005479.1 | 13, 14 | 1414 | |||
NR_024194.1 | 13, 14 | 1443 | ||||
NR_024195.1 | 12, 13 | 1364 | ||||
XM_011529818.2 | XP_011528120.1 | 14, 15 | 1428 | |||
XM_011529820.2 | XP_011528122.1 | 12, 13 | 1279 | |||
XM_017028529.1 | XP_016884018.1 | 14, 15 | 1428 | |||
XM_017028530.1 | XP_016884019.1 | 12, 13 | 1279 | |||
GenBank mRNA | AJ006973.1 | - | 13, 14 | 1350 | ||
AK090723.1 | - | 1, 2 | 923 | |||
AK097066.1 | - | 11, 12 | 925 | |||
AK097926.1 | - | 15, 16 | 1630 | |||
AK293694.1 | - | 13, 14 | 1341 | |||
AK295918.1 | - | 12, 13 | 1274 | |||
AK296670.1 | - | 13, 14 | 1351 | |||
AK297827.1 | - | 12, 13 | 1193 | |||
AK314522.1 | - | 13, 14 | 1323 | |||
BC040006.1 | - | 13, 14 | 1307 | |||
BC046151.1 | - | 13, 14 | 1292 | 6470390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
