siRNA Details
Gene Symbol: FAHD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016044.2 | NP_057128.2 | 8 | 1147 | ||
XM_011511289.2 | XP_011509591.1 | 8 | 1896 | |||
XM_017004236.1 | XP_016859725.1 | 9 | 1273 | |||
XM_017004237.1 | XP_016859726.1 | 9 | 1402 | |||
XM_017004238.1 | XP_016859727.1 | 9 | 1270 | |||
XM_017004239.1 | XP_016859728.1 | 8 | 1156 | |||
XM_017004240.1 | XP_016859729.1 | 8 | 1267 | |||
XM_017004243.1 | XP_016859732.1 | 7 | 1733 | |||
XM_017004244.1 | XP_016859733.1 | 7 | 993 | |||
GenBank mRNA | AF151863.1 | - | 8 | 1145 | ||
BC009403.2 | - | 8 | 1121 | 4309334 | ||
BC110911.1 | - | 8 | 1119 | 6064857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
