siRNA Details
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040138.2 | NP_001035228.1 | 1 | 208 | ||
NM_016326.3 | NP_057410.1 | 1 | 208 | |||
NM_016951.3 | NP_058647.1 | 1 | 208 | |||
NM_181640.2 | NP_857591.1 | 1 | 208 | |||
NM_181641.2 | NP_857592.1 | 1 | 208 | |||
GenBank mRNA | AA234074.1 | - | 1 | 132 | ||
AF057306.1 | - | 1 | 186 | |||
AF096895.2 | - | 1 | 202 | |||
AF135380.2 | - | 1 | 202 | |||
AF135381.2 | - | 1 | 202 | |||
AF145216.2 | - | 1 | 202 | |||
AF151058.1 | - | 1 | 205 | |||
AK290307.1 | - | 1 | 182 | |||
AK301240.1 | - | 1 | 199 | |||
AY358999.1 | - | 1 | 185 | |||
BC004380.1 | - | 1 | 191 | 3639550 | ||
BC091478.1 | - | 1 | 195 | 30333801 | ||
BU189908.1 | - | 1 | 168 | |||
DA113473.1 | - | 1 | 209 | |||
DA804889.1 | - | 1 | 189 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 1 | 208 | ||
NM_001204098.1 | NP_001191027.1 | 1 | 208 | |||
NM_001204099.1 | NP_001191028.1 | 1 | 208 | |||
GenBank mRNA | BM929895.1 | - | 1 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
