siRNA Details
Gene Symbol: SPIN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019003.3 | NP_061876.3 | 2 | 774 | ||
XM_005262016.4 | XP_005262073.3 | 2 | 427 | |||
XM_005262017.4 | XP_005262074.3 | 2 | 427 | |||
XM_005262019.3 | XP_005262076.1 | 2 | 1889 | |||
XM_017029598.1 | XP_016885087.1 | 2 | 1834 | |||
GenBank mRNA | BC071694.1 | - | 2 | 774 | 5201217 | |
Y09858.1 | - | 1 | 818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
