siRNA Details
Gene Symbol: CRCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019060.2 | NP_061933.1 | 2 | 531 | ||
XM_011509656.2 | XP_011507958.1 | 2 | 617 | |||
GenBank mRNA | AJ243662.1 | - | 2 | 531 | ||
BC119710.2 | - | 1 | 487 | 40117182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
