siRNA Details
Gene Symbol: ANGPTL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018687.6 | NP_061157.3 | 2 | 467 | ||
GenBank mRNA | AF271350.2 | - | 4 | 818 | ||
AY358356.1 | - | 2 | 451 | |||
BC113823.1 | - | 2 | 224 | |||
BP316755.1 | - | 2 | 468 | |||
KF809856.1 | - | 2 | 385 | |||
KF809857.1 | - | 2 | 239 |
