siRNA Details
Gene Symbol: PPAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020230.5 | NP_064615.3 | 12 | 1665 | ||
GenBank mRNA | AJ292529.1 | - | 12 | 1520 | ||
BC033202.1 | - | 12 | 1599 | 4871446 |
Gene Symbol: P2RY11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA310412.1 | - | 1 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
