siRNA Details
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 6, 7 | 791 | ||
NM_001005786.2 | NP_001005786.2 | 6, 7 | 791 | |||
NM_020363.3 | NP_065096.2 | 6, 7 | 791 | |||
GenBank mRNA | AF248480.1 | - | 6, 7 | 703 | ||
AF414184.1 | - | 6, 7 | 773 | |||
AK302398.1 | - | 6, 7 | 714 | |||
AK302690.1 | - | 5, 6 | 644 | |||
BC113006.1 | - | 6, 7 | 599 | 40035862 | ||
U21663.1 | - | 6, 7 | 715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
