siRNA Details
Gene Symbol: DLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286655.1 | NP_001273584.1 | 6 | 1859 | ||
NM_001286656.1 | NP_001273585.1 | 6 | 1536 | |||
NM_023932.3 | NP_076421.2 | 6 | 1554 | |||
NM_206539.2 | NP_996262.1 | 6 | 1970 | |||
XM_005249308.4 | XP_005249365.1 | 5 | 2762 | |||
XM_011514823.2 | XP_011513125.1 | 5 | 2123 | |||
XM_017011204.1 | XP_016866693.1 | 3 | 1572 | |||
GenBank mRNA | AK055380.1 | - | 6 | 1970 | ||
AY358126.1 | - | 6 | 1420 | |||
BC000230.1 | - | 6 | 1253 | 3350499 | ||
BC006425.1 | - | 6 | 1253 | 3349698 | ||
BC110320.1 | - | 6 | 1522 | 5495458 |
