siRNA Details
Gene Symbol: AHNAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001620.2 | NP_001611.1 | 4 | 477 | ||
NM_024060.3 | NP_076965.2 | 4 | 477 | |||
XM_005274240.1 | XP_005274297.1 | 4 | 374 | |||
XM_005274241.2 | XP_005274298.1 | 4 | 295 | |||
XM_005274245.1 | XP_005274302.1 | 4 | 419 | |||
XM_017018270.1 | XP_016873759.1 | 4 | 419 | |||
XM_017018271.1 | XP_016873760.1 | 4 | 419 | |||
XM_017018272.1 | XP_016873761.1 | 4 | 419 | |||
XM_017018273.1 | XP_016873762.1 | 4 | 419 | |||
GenBank mRNA | AK095346.1 | - | 4 | 418 | ||
AK300376.1 | - | 4 | 452 | |||
BC000926.1 | - | 4 | 419 | |||
BC012477.1 | - | 4 | 413 | |||
BC128460.1 | - | 4 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
