siRNA Details
Gene Symbol: HYI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190880.2 | NP_001177809.1 | 7, 8 | 957 | ||
XM_005271239.4 | XP_005271296.1 | 8, 9 | 1284 | |||
XM_005271240.4 | XP_005271297.1 | 7, 8 | 1182 | |||
GenBank mRNA | AF284750.1 | - | 7, 8 | 832 | ||
BC006140.1 | - | 7, 8 | 643 | 3928690 | ||
JF911821.1 | - | 6, 7 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
