siRNA Details
Gene Symbol: ABHD17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031213.3 | NP_112490.3 | 3, 4 | 884 | ||
XM_011528338.1 | XP_011526640.1 | 2, 3 | 3756 | |||
GenBank mRNA | BC020512.1 | - | 3, 4 | 798 | 3850505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
