siRNA Details
Gene Symbol: TBC1D3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291466.1 | NP_001278395.1 | 11 | 873 | ||
XM_006722254.1 | XP_006722317.1 | 14 | 1222 | |||
GenBank mRNA | AL136860.1 | - | 11 | 873 | ||
BC047739.1 | - | 12 | 1086 |
Gene Symbol: LOC101060389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291465.1 | NP_001278394.1 | 11 | 875 | ||
XM_006722248.2 | XP_006722311.1 | 10 | 2381 | |||
XM_006722250.2 | XP_006722313.1 | 10 | 2381 | |||
XM_006722253.2 | XP_006722316.1 | 10 | 2381 | |||
XM_011524176.2 | XP_011522478.1 | 11 | 988 | |||
GenBank mRNA | BI088323.1 | - | 9 | 638 |
Gene Symbol: TBC1D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123391.3 | NP_001116863.3 | 11 | 887 | ||
GenBank mRNA | AB449884.1 | - | 10 | 775 | ||
AK302064.1 | - | 12 | 1113 | |||
AK303259.1 | - | 10 | 907 | |||
AX775941.1 | - | 13 | 1136 | |||
AX775943.1 | - | 13 | 1136 |
Gene Symbol: TBC1D3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001417.6 | NP_001001417.6 | 11 | 921 | ||
XM_005257980.4 | XP_005258037.2 | 11 | 1051 | |||
GenBank mRNA | AF540953.1 | - | 10 | 775 | ||
AK292257.1 | - | 11 | 892 |
Gene Symbol: TBC1D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001418.6 | NP_001001418.5 | 11 | 915 | ||
XM_006721904.2 | XP_006721967.1 | 10 | 2381 | |||
XM_006721908.2 | XP_006721971.1 | 10 | 2381 | |||
XM_011524814.2 | XP_011523116.1 | 11 | 988 | |||
XM_017024646.1 | XP_016880135.1 | 10 | 2381 | |||
GenBank mRNA | AK313329.1 | - | 11 | 915 |
Gene Symbol: TBC1D3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032258.4 | NP_115634.3 | 11 | 889 | ||
GenBank mRNA | AK225985.1 | - | 11 | 880 | ||
AK292344.1 | - | 11 | 932 | |||
BC075809.1 | - | 11 | 921 | 5170064 |
Gene Symbol: TBC1D3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291462.1 | NP_001278391.1 | 11 | 875 | ||
XM_005276914.3 | XP_005276971.1 | 11 | 2700 | |||
GenBank mRNA | AK292308.1 | - | 12 | 925 | ||
AK301823.1 | - | 9 | 687 | |||
BC071680.1 | - | 11 | 875 | 5165385 |
Gene Symbol: TBC1D3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123392.3 | NP_001116864.3 | 11 | 889 | ||
XM_006722055.1 | XP_006722118.1 | 10 | 1529 | |||
XM_011525172.1 | XP_011523474.1 | 10 | 1712 | |||
XM_011525173.2 | XP_011523475.1 | 11 | 1083 | |||
XM_011525175.1 | XP_011523477.1 | 10 | 1712 | |||
XM_011525176.1 | XP_011523478.1 | 10 | 1712 | |||
GenBank mRNA | BM844606.1 | - | 2 | 87 | ||
BX281608.1 | - | 5 | 335 |
Gene Symbol: TBC1D3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291463.1 | NP_001278392.1 | 11 | 875 | ||
XM_006722223.3 | XP_006722286.1 | 11 | 1146 | |||
XM_006722224.3 | XP_006722287.1 | 10 | 2391 | |||
XM_011524184.2 | XP_011522486.1 | 10 | 2574 | |||
XM_011524185.2 | XP_011522487.1 | 10 | 2574 |
Gene Symbol: TBC1D3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025494.1 | XP_016880983.1 | 9 | 691 |
Gene Symbol: TBC1D3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291464.1 | NP_001278393.1 | 11 | 875 | ||
XM_006722235.2 | XP_006722298.1 | 10 | 3153 | |||
XM_006722236.1 | XP_006722299.1 | 11 | 1725 | |||
XM_006722237.2 | XP_006722300.1 | 10 | 2970 | |||
XM_011524173.1 | XP_011522475.1 | 10 | 3153 | |||
XM_011524174.1 | XP_011522476.1 | 10 | 3153 |
Gene Symbol: TBC1D3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291459.1 | NP_001278388.1 | 11 | 889 | ||
XM_006722242.3 | XP_006722305.1 | 10 | 4129 | |||
XM_006722244.2 | XP_006722307.1 | 10 | 2381 | |||
XM_006722247.2 | XP_006722310.1 | 10 | 2381 | |||
XM_011524175.2 | XP_011522477.1 | 11 | 988 | |||
GenBank mRNA | AK122833.1 | - | 7 | 2200 |
Gene Symbol: TBC1D3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027486.1 | 11 | 834 | |||
GenBank mRNA | AK301746.1 | - | 11 | 835 | ||
BC058890.1 | - | 11 | 859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
