siRNA Details
Gene Symbol: TBC1D3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123392.3 | NP_001116864.3 | 14 | 1761 | ||
XM_006722055.1 | XP_006722118.1 | 13 | 2401 | |||
XM_006722056.1 | XP_006722119.1 | 12 | 2335 | |||
XM_011525172.1 | XP_011523474.1 | 13 | 2584 | |||
XM_011525173.2 | XP_011523475.1 | 14 | 1955 | |||
XM_011525174.1 | XP_011523476.1 | 12 | 2518 | |||
XM_011525175.1 | XP_011523477.1 | 12 | 2431 | |||
GenBank mRNA | BI597531.1 | - | 2 | 656 | ||
DB512851.2 | - | 1 | 347 |
Gene Symbol: LOC101060389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291465.1 | NP_001278394.1 | 14 | 1747 | ||
XM_006722248.2 | XP_006722311.1 | 13 | 3253 | |||
XM_006722249.2 | XP_006722312.1 | 12 | 3187 | |||
XM_006722250.2 | XP_006722313.1 | 12 | 3100 | |||
XM_011524176.2 | XP_011522478.1 | 14 | 1860 |
Gene Symbol: TBC1D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123391.3 | NP_001116863.3 | 14 | 1759 | ||
GenBank mRNA | AK302064.1 | - | 15 | 1985 | ||
AK303259.1 | - | 13 | 1779 | |||
AX775941.1 | - | 17 | 1986 | |||
AX775943.1 | - | 17 | 1986 |
Gene Symbol: TBC1D3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001417.6 | NP_001001417.6 | 14 | 1793 | ||
XM_005257980.4 | XP_005258037.2 | 14 | 1923 | |||
GenBank mRNA | AF540953.1 | - | 13 | 1647 | ||
AK292257.1 | - | 14 | 1764 |
Gene Symbol: TBC1D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001418.6 | NP_001001418.5 | 14 | 1787 | ||
XM_006721904.2 | XP_006721967.1 | 13 | 3253 | |||
XM_006721905.2 | XP_006721968.1 | 12 | 3187 | |||
XM_011524814.2 | XP_011523116.1 | 14 | 1860 | |||
XM_017024646.1 | XP_016880135.1 | 12 | 3100 |
Gene Symbol: TBC1D3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291466.1 | NP_001278395.1 | 14 | 1745 | ||
XM_006722254.1 | XP_006722317.1 | 17 | 2094 | |||
GenBank mRNA | AL136860.1 | - | 14 | 1745 | ||
BC047739.1 | - | 15 | 2151 |
Gene Symbol: TBC1D3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032258.4 | NP_115634.3 | 14 | 1761 | ||
GenBank mRNA | AK225985.1 | - | 14 | 1752 | ||
AK292344.1 | - | 14 | 1804 | |||
BC075809.1 | - | 14 | 1793 | 5170064 |
Gene Symbol: TBC1D3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291462.1 | NP_001278391.1 | 14 | 1747 | ||
XM_005276914.3 | XP_005276971.1 | 14 | 3572 | |||
GenBank mRNA | AK292308.1 | - | 15 | 1797 | ||
AK301823.1 | - | 12 | 1559 | |||
BC071680.1 | - | 14 | 1747 | 5165385 |
Gene Symbol: TBC1D3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291463.1 | NP_001278392.1 | 14 | 1747 | ||
XM_006722223.3 | XP_006722286.1 | 14 | 2018 | |||
XM_006722224.3 | XP_006722287.1 | 13 | 3263 | |||
XM_006722226.3 | XP_006722289.1 | 12 | 3197 | |||
XM_011524183.2 | XP_011522485.1 | 12 | 3380 | |||
XM_011524184.2 | XP_011522486.1 | 12 | 3293 | |||
GenBank mRNA | BC078140.1 | - | 12 | 1582 | 5742850 |
Gene Symbol: TBC1D3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291464.1 | NP_001278393.1 | 14 | 1747 | ||
XM_006722235.2 | XP_006722298.1 | 13 | 4025 | |||
XM_006722236.1 | XP_006722299.1 | 14 | 2597 | |||
XM_006722237.2 | XP_006722300.1 | 13 | 3842 | |||
XM_006722239.2 | XP_006722302.1 | 12 | 3776 | |||
XM_011524172.1 | XP_011522474.1 | 12 | 3959 | |||
XM_011524173.1 | XP_011522475.1 | 12 | 3872 |
Gene Symbol: TBC1D3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291459.1 | NP_001278388.1 | 14 | 1761 | ||
XM_006722242.3 | XP_006722305.1 | 13 | 5001 | |||
XM_006722243.2 | XP_006722306.1 | 12 | 3187 | |||
XM_006722244.2 | XP_006722307.1 | 12 | 3100 | |||
XM_006722246.3 | XP_006722309.1 | 12 | 2335 | |||
XM_011524175.2 | XP_011522477.1 | 14 | 1860 | |||
GenBank mRNA | AK095385.1 | - | 12 | 2335 | ||
AK122833.1 | - | 10 | 3072 | |||
AX775949.1 | - | 12 | 2335 | |||
AX775951.1 | - | 12 | 2335 |
Gene Symbol: TBC1D3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027486.1 | 14 | 1748 | |||
GenBank mRNA | BC058890.1 | - | 14 | 1773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
