siRNA Details
Gene Symbol: ZDHHC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318015.1 | NP_001304944.1 | 2 | 415 | ||
NM_001318016.1 | NP_001304945.1 | 2 | 250 | |||
NM_001318020.1 | NP_001304949.1 | 2 | 250 | |||
NM_001318023.1 | NP_001304952.1 | 2 | 250 | |||
NM_032799.4 | NP_116188.2 | 2 | 250 | |||
XM_011519116.2 | XP_011517418.1 | 2 | 424 | |||
GenBank mRNA | AK027430.1 | - | 2 | 243 | ||
AK075332.1 | - | 2 | 243 | |||
AK092843.1 | - | 1 | 1667 | |||
AK315746.1 | - | 2 | 246 | |||
BC009280.2 | - | 2 | 227 | 4302257 | ||
BC048251.1 | - | 2 | 396 | 5418875 | ||
BG767826.1 | - | 2 | 224 | |||
BM043458.1 | - | 2 | 218 | |||
BM745076.1 | - | 2 | 242 | |||
BU506687.1 | - | 2 | 251 |
