siRNA Details
Gene Symbol: ZNF251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138367.1 | NP_612376.1 | 4 | 477 | ||
XM_005272356.4 | XP_005272413.1 | 4 | 495 | |||
XM_006716683.2 | XP_006716746.1 | 3 | 296 | |||
XM_011517361.1 | XP_011515663.1 | 3 | 296 | |||
GenBank mRNA | AK000435.1 | - | 4 | 361 | ||
AK091638.1 | - | 2 | 2284 | |||
DA811573.1 | - | 4 | 477 |
