siRNA Details
Gene Symbol: ZNF837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138466.1 | NP_612475.1 | 3 | 1900 | ||
NR_049780.1 | 3 | 2002 | ||||
XM_017026242.1 | XP_016881731.1 | 2 | 2668 | |||
GenBank mRNA | BC012365.1 | - | 2 | 1762 | 4576341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
