siRNA Details
Gene Symbol: OVCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080822.2 | NP_543012.1 | 1, 2 | 243 | ||
GenBank mRNA | AF073528.1 | - | 1, 2 | 178 | ||
AF321875.1 | - | 1, 2 | 178 | |||
BC040696.2 | - | 1, 2 | 193 | 6158258 | ||
BC041170.1 | - | 1, 2 | 193 | 5738727 | ||
CB137429.1 | - | 1, 2 | 243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
