siRNA Details
Gene Symbol: NTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145807.2 | NP_665806.1 | 7 | 1674 | ||
XM_017026274.1 | XP_016881763.1 | 6 | 1900 | |||
GenBank mRNA | BC018654.2 | - | 7 | 1600 | 2961115 | |
BC018697.2 | - | 7 | 1600 | 2961115 | ||
BC033207.1 | - | 7 | 1600 | 4903484 |
