siRNA Details
Gene Symbol: C1orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122961.1 | NP_001116433.1 | 3 | 404 | ||
XM_005270448.3 | XP_005270505.1 | 2 | 307 | |||
XM_006710342.3 | XP_006710405.1 | 3 | 750 | |||
XM_006710343.3 | XP_006710406.1 | 2 | 633 | |||
XM_011540647.2 | XP_011538949.1 | 3 | 781 | |||
GenBank mRNA | BG196327.1 | - | 1 | 67 | ||
BG772797.1 | - | 3 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
