siRNA Details
Gene Symbol: BHLHE23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080606.3 | NP_542173.2 | 1 | 758 | ||
GenBank mRNA | BC137288.1 | - | 1 | 758 | 9021285 | |
BC137308.1 | - | 1 | 758 | 9021305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
