siRNA Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 3 | 1789 | |||
NR_026926.1 | 3 | 676 | ||||
GenBank mRNA | AF451986.1 | - | 2 | 267 | ||
AK057399.1 | - | 3 | 1723 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 3 | 1789 | |||
NR_026926.1 | 3 | 676 | ||||
GenBank mRNA | AF451986.1 | - | 2 | 267 | ||
AK057399.1 | - | 3 | 1723 |