siRNA Details
Gene Symbol: CSAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102576.2 | NP_001096046.2 | 3 | 265 | ||
NM_153478.2 | NP_705611.2 | 4 | 481 | |||
GenBank mRNA | AF195880.1 | - | 4 | 479 | ||
AF268418.1 | - | 3 | 235 | |||
AF268419.1 | - | 4 | 432 | |||
BC059947.1 | - | 3 | 235 | 5397842 | ||
BF570999.1 | - | 3 | 264 | |||
BM810082.1 | - | 3 | 247 |
