siRNA Details
Gene Symbol: PRR22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134316.1 | NP_001127788.1 | 3 | 1156 | ||
GenBank mRNA | BC008985.1 | - | 1 | 1404 | 4213313 | |
BC023278.1 | - | 1 | 1404 | 4213314 |
