siRNA Details
Gene Symbol: PRR22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008985.1 | - | 1 | 546 | 4213313 | |
BC023278.1 | - | 1 | 546 | 4213314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
